Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001193360.1 | NP_001180289.1 | 9 | 1830 | |
NM_001193361.1 | NP_001180290.1 | 8 | 1746 | ||
NM_001193362.1 | NP_001180291.1 | 9 | 1989 | ||
NM_001193363.1 | NP_001180292.1 | 8 | 1905 | ||
NM_018199.3 | NP_060669.1 | 7 | 1366 | ||
NR_034164.1 | 10 | 1920 | |||
NR_034165.1 | 10 | 2079 | |||
XM_005267817.3 | XP_005267874.1 | 7 | 1296 | ||
XM_011536908.2 | XP_011535210.1 | 7 | 1384 | ||
XM_017021421.1 | XP_016876910.1 | 8 | 1429 | ||
GenBank mRNA | AK001600.1 | - | 7 | 1294 | |
AK056025.1 | - | 10 | 1866 | ||
AK292563.1 | - | 9 | 1989 | ||
AK295601.1 | - | 8 | 1865 | ||
AL834434.1 | - | 7 | 1288 | ||
BC001962.1 | - | 7 | 1426 | 3537432 | |
BX647767.1 | - | 8 | 1696 | ||
CR936728.1 | - | 9 | 1705 | ||
CR936729.1 | - | 8 | 1713 |