Gene Transcripts

Gene Symbol
LRRN3
Entrez Gene ID
54674
Gene Name
leucine rich repeat neuronal 3
Gene Aliases
FIGLER5, NLRR-3, NLRR3
Chromosome Location
Chr. 7: 111091006 - 111125454 on Build GRCh38
UniGene ID
Hs.3781
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099658.1 NP_001093128.1 3 1422
NM_001099660.1 NP_001093130.1 4 1529
NM_018334.4 NP_060804.3 2 1340
GenBank mRNA AB060967.1 - 1 789
AF134481.1 - 1 820
AK001991.1 - 4 1441
AK292556.1 - 2 929
AL442092.1 - 1 745
AY358280.1 - 3 962
BC035133.2 - 2 1219 5263900
CR457397.1 - 1 483

View Full Product Details