Gene Transcripts

Gene Symbol
KIZ
Entrez Gene ID
55857
Gene Name
kizuna centrosomal protein
Gene Aliases
C20orf19, HT013, Kizuna, NCRNA00153, PLK1S1, RP69
Chromosome Location
Chr. 20: 21125980 - 21246628 on Build GRCh38
UniGene ID
Hs.187635
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018474.4 NP_060944.3 3 387
XM_011529296.2 XP_011527598.1 3 390
XM_011529297.2 XP_011527599.1 3 379
XM_011529298.2 XP_011527600.1 3 372
XM_017027951.1 XP_016883440.1 3 289
XM_017027953.1 XP_016883442.1 3 361
GenBank mRNA AF451990.1 - 3 365
AK025476.1 - 3 318
BC039296.1 - 3 349
BC105093.1 - 3 290 8144096
BC113370.1 - 3 290 8322422

View Full Product Details