Gene Transcripts

Gene Symbol
PCDHB7
Entrez Gene ID
56129
Gene Name
protocadherin beta 7
Gene Aliases
PCDH-BETA7
Chromosome Location
Chr. 5: 141172621 - 141176376 on Build GRCh38
UniGene ID
Hs.203830
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018940.3 NP_061763.1 1 1246
GenBank mRNA AF152500.1 - 1 1031
AF217750.1 - 1 1031
AK024641.1 - 1 1205
AK222560.1 - 1 1207
AK222678.1 - 1 1191
AK315054.1 - 1 1249
AL834254.1 - 1 593
BC098574.1 - 1 1185 30925247
BC130331.1 - 1 1113 40146362
BC130333.1 - 1 1113 40146364

View Full Product Details