Gene Transcripts
- Gene Symbol
- SNTG2
- Entrez Gene ID
- 54221
- Gene Name
- syntrophin gamma 2
- Gene Aliases
- G2SYN, SYN5
- Chromosome Location
- Chr. 2: 950868 - 1367991 on Build GRCh38
- UniGene ID
- Hs.595069
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018968.3 |
NP_061841.2
|
5, 6 |
506 |
|
XM_011510366.2 |
XP_011508668.1
|
3, 4 |
543 |
|
XM_017004361.1 |
XP_016859850.1
|
5, 6 |
468 |
|
XM_017004362.1 |
XP_016859851.1
|
5, 6 |
474 |
|
XM_017004363.1 |
XP_016859852.1
|
5, 6 |
655 |
|
XM_017004364.1 |
XP_016859853.1
|
2, 3 |
3796 |
|
XM_017004365.1 |
XP_016859854.1
|
5, 6 |
469 |
|
XM_017004366.1 |
XP_016859855.1
|
5, 6 |
469 |
|
XM_017004367.1 |
XP_016859856.1
|
6, 7 |
544 |
|
XM_017004368.1 |
XP_016859857.1
|
5, 6 |
469 |
|
XM_017004369.1 |
XP_016859858.1
|
5, 6 |
469 |
|
XM_017004370.1 |
XP_016859859.1
|
5, 6 |
469 |
|
XM_017004371.1 |
XP_016859860.1
|
5, 6 |
470 |
|
XM_017004372.1 |
XP_016859861.1
|
5, 6 |
470 |
|
XM_017004374.1 |
XP_016859863.1
|
5, 6 |
475 |
|
GenBank mRNA |
AJ003029.1 |
- |
5, 6 |
508 |
|
View Full Product Details