Gene Transcripts

Gene Symbol
UBQLN4
Entrez Gene ID
56893
Gene Name
ubiquilin 4
Gene Aliases
A1U, A1Up, C1orf6, CIP75, UBIN
Chromosome Location
Chr. 1: 156035294 - 156053825 on Build GRCh38
UniGene ID
Hs.283739
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304342.1 NP_001291271.1 11 2103
NM_020131.4 NP_064516.2 11 2163
GenBank mRNA AB209566.1 - 11 2036
AF113544.1 - 8 1568
AF188240.1 - 11 2317
AK098368.1 - 6 1008
BC006410.1 - 2 438
BC018403.2 - 11 2051 4214386
BC063841.1 - 11 2063 6183942

View Full Product Details