Gene Transcripts

Gene Symbol
THAP10
Entrez Gene ID
56906
Gene Name
THAP domain containing 10
Gene Aliases
Chromosome Location
Chr. 15: 70881342 - 70892433 on Build GRCh38
UniGene ID
Hs.591123
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020147.3 NP_064532.1 3 1409
GenBank mRNA AL360202.1 - 3 1368
BC027857.1 - 3 1390 5222191
BC072414.1 - 3 1397 6167441

View Full Product Details