Gene Transcripts

Gene Symbol
HIST1H2AB
Entrez Gene ID
8335
Gene Name
histone cluster 1, H2ab
Gene Aliases
H2A/m, H2AFM
Chromosome Location
Chr. 6: 26033092 - 26033568 on Build GRCh38
UniGene ID
Hs.248174
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003513.2 NP_003504.2 1 386
GenBank mRNA BC125140.1 - 1 390

View Full Product Details