Gene Transcripts
- Gene Symbol
- SNX15
- Entrez Gene ID
- 29907
- Gene Name
- sorting nexin 15
- Gene Aliases
- HSAF001435
- Chromosome Location
- Chr. 11: 65027408 - 65040572 on Build GRCh38
- UniGene ID
- Hs.80132
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013306.4 |
NP_037438.2
|
1, 2 |
230 |
|
NM_147777.3 |
NP_680086.2
|
1, 2 |
230 |
|
GenBank mRNA |
AB209439.1 |
- |
1, 2 |
98 |
|
AF001435.1 |
- |
1, 2 |
156 |
|
AF175267.2 |
- |
1, 2 |
139 |
|
AF175268.1 |
- |
1, 2 |
100 |
|
AK315034.1 |
- |
1, 2 |
161 |
|
AK315642.1 |
- |
1, 2 |
171 |
|
BC009897.2 |
- |
1, 2 |
148 |
3510379 |
BC012767.2 |
- |
1, 2 |
200 |
3678551 |
BC014520.2 |
- |
1, 2 |
200 |
3678576 |
BT006631.1 |
- |
1, 2 |
100 |
|
DA079695.1 |
- |
1, 2 |
231 |
|
- Gene Symbol
- ARL2-SNX15
- Entrez Gene ID
- 100528018
- Gene Name
- ARL2-SNX15 readthrough (NMD candidate)
- Gene Aliases
- Chromosome Location
- Chr. 11: 65014113 - 65040572 on Build GRCh38
- UniGene ID
- Hs.80132
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037650.1 |
|
4, 5 |
754 |
|
GenBank mRNA |
AK096447.1 |
- |
4, 5 |
688 |
|
View Full Product Details