Gene Transcripts

Gene Symbol
VENTX
Entrez Gene ID
27287
Gene Name
VENT homeobox
Gene Aliases
HPX42B, NA88A, VENTX2
Chromosome Location
Chr. 10: 133237904 - 133241930 on Build GRCh38
UniGene ID
Hs.125231
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014468.3 NP_055283.1 3 1326
GenBank mRNA AF068006.1 - 3 1333

View Full Product Details