Gene Transcripts

Gene Symbol
SCHIP1
Entrez Gene ID
29970
Gene Name
schwannomin interacting protein 1
Gene Aliases
SCHIP-1
Chromosome Location
Chr. 3: 159273247 - 159897366 on Build GRCh38
UniGene ID
Hs.134665
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001197107.1 NP_001184036.1 6 1797
NM_001197108.1 NP_001184037.1 5 1140
NM_001197109.1 NP_001184038.1 5 857
NM_014575.3 NP_055390.1 6 1836
GenBank mRNA AF070614.1 - 5 946
AF145713.1 - 6 1331
AF145714.1 - 5 635
AF145715.1 - 6 1292
AK091871.1 - 5 778
AK309223.1 - 6 1244
AK314662.1 - 6 1779
BC005947.1 - 5 673 4246326
BC036535.2 - 6 1836 5263291
BX648179.1 - 3 3268

Gene Symbol
IQCJ-SCHIP1
Entrez Gene ID
100505385
Gene Name
IQCJ-SCHIP1 readthrough
Gene Aliases
Chromosome Location
Chr. 3: 159069252 - 159897366 on Build GRCh38
UniGene ID
Hs.134665
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001197113.1 NP_001184042.1 9 1671
NM_001197114.1 NP_001184043.1 8 1590
GenBank mRNA AK096479.1 - 9 1577
DQ157847.1 - 7 1409
DQ157848.1 - 8 1490

View Full Product Details