Gene Transcripts

Gene Symbol
MRPL27
Entrez Gene ID
51264
Gene Name
mitochondrial ribosomal protein L27
Gene Aliases
L27mt
Chromosome Location
Chr. 17: 50367867 - 50373201 on Build GRCh38
UniGene ID
Hs.7736
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016504.2 NP_057588.1 4 693
GenBank mRNA AF151084.1 - 4 677
BG829493.1 - 4 712

View Full Product Details