Gene Transcripts

Gene Symbol
BLCAP
Entrez Gene ID
10904
Gene Name
bladder cancer associated protein
Gene Aliases
BC10
Chromosome Location
Chr. 20: 37517417 - 37527931 on Build GRCh38
UniGene ID
Hs.472651
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167820.1 NP_001161292.1 3 1684
NM_001167821.1 NP_001161293.1 2 1514
NM_001167822.2 NP_001161294.1 2 1571
NM_001167823.1 NP_001161295.1 2 1587
NM_001317074.1 NP_001304003.1 2 1564
NM_001317075.1 NP_001304004.1 2 1480
NM_006698.3 NP_006689.1 2 1551
XM_017027599.1 XP_016883088.1 2 2464
GenBank mRNA AF053470.1 - 2 1492
AK074498.1 - 5 1300
AK124822.1 - 3 1411
AL049288.1 - 2 1497
BC047692.1 - 2 1482 6059195
BC071704.1 - 2 1476 5190373

View Full Product Details