Gene Transcripts

Gene Symbol
FERMT2
Entrez Gene ID
10979
Gene Name
fermitin family member 2
Gene Aliases
KIND2, MIG2, PLEKHC1, UNC112, UNC112B, mig-2
Chromosome Location
Chr. 14: 52857271 - 52951192 on Build GRCh38
UniGene ID
Hs.509343
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134999.1 NP_001128471.1 3 427
NM_001135000.1 NP_001128472.1 3 427
NM_006832.2 NP_006823.1 3 427
XM_005267285.2 XP_005267342.1 3 522
XM_006720008.2 XP_006720071.1 3 522
XM_006720009.2 XP_006720072.1 3 522
XM_006720010.2 XP_006720073.1 3 522
GenBank mRNA AF443279.1 - 3 322
AK291738.1 - 3 380
BC017327.2 - 3 427 4547604
BX161467.1 - 3 348
EU979385.1 - 2 241
Z24725.1 - 3 363

View Full Product Details