Gene Transcripts

Gene Symbol
VASH1
Entrez Gene ID
22846
Gene Name
vasohibin 1
Gene Aliases
KIAA1036
Chromosome Location
Chr. 14: 76761457 - 76783020 on Build GRCh38
UniGene ID
Hs.525479
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014909.4 NP_055724.1 7 3782
XM_017021087.1 XP_016876576.1 9 2857
XM_017021088.1 XP_016876577.1 8 2814
XM_017021089.1 XP_016876578.1 8 2673
XM_017021090.1 XP_016876579.1 7 2606
XM_017021091.1 XP_016876580.1 7 4282
GenBank mRNA AB028959.1 - 7 3237
AL832588.2 - 7 3485

View Full Product Details