Gene Transcripts

Gene Symbol
COLGALT2
Entrez Gene ID
23127
Gene Name
collagen beta(1-O)galactosyltransferase 2
Gene Aliases
C1orf17, GLT25D2
Chromosome Location
Chr. 1: 183929662 - 184051612 on Build GRCh38
UniGene ID
Hs.387995
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001303420.1 NP_001290349.1 3 888
NM_001303421.1 NP_001290350.1 3 402
NM_015101.3 NP_055916.1 3 888
XM_011509332.2 XP_011507634.1 3 595
XM_011509334.2 XP_011507636.1 3 402
XM_011509335.2 XP_011507637.1 3 453
XM_011509336.2 XP_011507638.1 3 230
XM_017000750.1 XP_016856239.1 3 453
GenBank mRNA AB011156.1 - 3 813
AF288389.1 - 3 847
AK095192.1 - 3 376
AK293973.1 - 3 595
BC035672.1 - 3 870 5740842
DA216863.1 - 3 402

View Full Product Details