Gene Transcripts

Gene Symbol
WDR7
Entrez Gene ID
23335
Gene Name
WD repeat domain 7
Gene Aliases
TRAG
Chromosome Location
Chr. 18: 56651343 - 57029805 on Build GRCh38
UniGene ID
Hs.465213
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015285.2 NP_056100.2 2 339
NM_052834.2 NP_443066.2 2 339
XM_006722431.2 XP_006722494.1 2 271
XM_011525888.1 XP_011524190.1 2 381
XM_017025682.1 XP_016881171.1 2 244
GenBank mRNA AB011113.2 - 2 302
AF188125.1 - 1 128
AK057141.1 - 2 363
AY099325.1 - 1 128
BC131558.1 - 1 128 40108655
BC150282.1 - 2 302 9007268

View Full Product Details