Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001297576.1 | NP_001284505.1 | 6 | 938 | |
NM_001297577.1 | NP_001284506.1 | 5 | 736 | ||
NM_001297578.1 | NP_001284507.1 | 4 | 565 | ||
NM_003768.4 | NP_003759.1 | 4 | 631 | ||
NR_123724.1 | 3 | 457 | |||
XM_006711599.2 | XP_006711662.1 | 4 | 535 | ||
GenBank mRNA | AK095879.1 | - | 6 | 843 | |
BC002426.2 | - | 4 | 507 | 3346270 | |
BC022554.1 | - | 4 | 618 | 4820225 | |
DA157938.1 | - | 4 | 501 | ||
DA538845.1 | - | 3 | 395 | ||
X86809.1 | - | 4 | 538 | ||
Y13736.1 | - | 4 | 494 |