Gene Transcripts

Gene Symbol
LY86
Entrez Gene ID
9450
Gene Name
lymphocyte antigen 86
Gene Aliases
MD-1, MD1, MMD-1, dJ80N2.1
Chromosome Location
Chr. 6: 6588639 - 6654983 on Build GRCh38
UniGene ID
Hs.653138
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004271.3 NP_004262.1 3 294
XM_017011505.1 XP_016866994.1 3 356
GenBank mRNA AB020499.1 - 2 130
AF057178.2 - 3 283
AY931019.1 - 3 260
BC038846.1 - 3 304 6015123
CD695164.1 - 3 358

View Full Product Details