Gene Transcripts

Gene Symbol
SLK
Entrez Gene ID
9748
Gene Name
STE20 like kinase
Gene Aliases
LOSK, STK2, bA16H23.1, se20-9
Chromosome Location
Chr. 10: 103967173 - 104029233 on Build GRCh38
UniGene ID
Hs.591922
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014720.3 NP_055535.2 12, 13 3350
XM_011540401.2 XP_011538703.1 11, 12 2006
GenBank mRNA AF273048.1 - 12, 13 2823

View Full Product Details