Gene Transcripts

Gene Symbol
SSPN
Entrez Gene ID
8082
Gene Name
sarcospan
Gene Aliases
DAGA5, KRAG, NSPN, SPN1, SPN2
Chromosome Location
Chr. 12: 26122024 - 26234777 on Build GRCh38
UniGene ID
Hs.183428
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135823.1 NP_001129295.1 3 1487
NM_005086.4 NP_005077.2 3 1670
XM_011520853.2 XP_011519155.1 3 1420
XM_011520854.2 XP_011519156.1 3 1594
XM_011520855.2 XP_011519157.1 3 1336
GenBank mRNA AK092925.1 - 3 1483
AL136756.1 - 3 1654

View Full Product Details