Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001297575.1 | NP_001284504.1 | 7 | 1428 | |
NM_014625.3 | NP_055440.1 | 8 | 1632 | ||
XM_005245483.3 | XP_005245540.1 | 6 | 1477 | ||
XM_017002298.1 | XP_016857787.1 | 5 | 1329 | ||
GenBank mRNA | AJ279254.1 | - | 8 | 1613 | |
BC029141.1 | - | 7 | 1428 | 5187188 | |
BX095913.1 | - | 1 | 388 |