Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290230.1 | NP_001277159.1 | 5 | 1151 | |
NM_001798.4 | NP_001789.2 | 7 | 1331 | ||
NM_052827.3 | NP_439892.2 | 6 | 1229 | ||
XM_011537732.1 | XP_011536034.1 | 8 | 1475 | ||
GenBank mRNA | AK291941.1 | - | 7 | 1270 | |
BC003065.2 | - | 7 | 1251 | 3504348 | |
M68520.1 | - | 7 | 1281 | ||
X61622.1 | - | 7 | 1093 | ||
X62071.1 | - | 7 | 1261 |