Gene Transcripts

Gene Symbol
LYSMD4
Entrez Gene ID
145748
Gene Name
LysM domain containing 4
Gene Aliases
Chromosome Location
Chr. 15: 99715033 - 99733561 on Build GRCh38
UniGene ID
Hs.562568
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284419.1 NP_001271348.1 3 422
NM_152449.3 NP_689662.2 4 537
XM_011521244.1 XP_011519546.1 2 725
XM_017021927.1 XP_016877416.1 2 725
XM_017021929.1 XP_016877418.1 2 725
XM_017021930.1 XP_016877419.1 2 739
GenBank mRNA AK124852.1 - 2 675
BC041097.1 - 4 514 5240362
DA405564.1 - 3 415

View Full Product Details