Gene Transcripts
- Gene Symbol
- CFAP161
- Entrez Gene ID
- 161502
- Gene Name
- cilia and flagella associated protein 161
- Gene Aliases
- C15orf26
- Chromosome Location
- Chr. 15: 81099408 - 81149175 on Build GRCh38
- UniGene ID
- Hs.130979
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK095934.1 |
- |
8 |
1215 |
|
View Full Product Details