Gene Transcripts

Gene Symbol
SMARCC1
Entrez Gene ID
6599
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 1
Gene Aliases
BAF155, CRACC1, Rsc8, SRG3, SWI3
Chromosome Location
Chr. 3: 47585888 - 47781915 on Build GRCh38
UniGene ID
Hs.476179
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003074.3 NP_003065.3 5, 6 702
XM_011534034.1 XP_011532336.1 5, 6 433
GenBank mRNA AK302541.1 - 5, 6 477
BC021862.1 - 5, 6 639
BC032564.1 - 5, 6 686
BC039843.1 - 5, 6 606
BC050564.1 - 5, 6 631 3912713
BC065253.1 - 5, 6 671
BC113465.1 - 5, 6 650 8322517
BC117213.1 - 5, 6 650 40125764
U66615.1 - 5, 6 638

View Full Product Details