Gene Transcripts

Gene Symbol
RBBP4
Entrez Gene ID
5928
Gene Name
RB binding protein 4, chromatin remodeling factor
Gene Aliases
NURF55, RBAP48, lin-53
Chromosome Location
Chr. 1: 32651148 - 32686211 on Build GRCh38
UniGene ID
Hs.16003
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135255.1 NP_001128727.1 12 1659
NM_001135256.1 NP_001128728.1 12 1562
NM_005610.2 NP_005601.1 12 1662
GenBank mRNA AK222779.1 - 12 1565
AK299251.1 - 12 1562
AK299410.1 - 10 1273
BC003092.1 - 12 1589 3356558
BC015123.1 - 10 1323
BC053904.1 - 12 1575 6526200
BC075836.1 - 12 1530 6338602
X74262.1 - 12 1586

View Full Product Details