Gene Transcripts

Gene Symbol
PRKCQ
Entrez Gene ID
5588
Gene Name
protein kinase C theta
Gene Aliases
PRKCT, nPKC-theta
Chromosome Location
Chr. 10: 6393038 - 6580646 on Build GRCh38
UniGene ID
Hs.498570
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242413.2 NP_001229342.1 17 2505
NM_001282644.1 NP_001269573.1 18 2692
NM_001282645.1 NP_001269574.1 17 2633
NM_001323265.1 NP_001310194.1 18 2749
NM_001323266.1 NP_001310195.1 17 2631
NM_006257.4 NP_006248.1 18 2694
XM_005252496.4 XP_005252553.1 18 2794
XM_006717465.3 XP_006717528.1 20 5189
XM_017016410.1 XP_016871899.1 19 5100
XM_017016411.1 XP_016871900.1 19 5128
GenBank mRNA BX647657.1 - 17 2779
L01087.1 - 18 2688

View Full Product Details