Gene Transcripts

Gene Symbol
MFAP1
Entrez Gene ID
4236
Gene Name
microfibrillar associated protein 1
Gene Aliases
Chromosome Location
Chr. 15: 43804535 - 43824753 on Build GRCh38
UniGene ID
Hs.61418
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005926.2 NP_005917.2 9 1358
GenBank mRNA AK312867.1 - 9 1334
BC023557.2 - 9 1293 4139958
BC050742.1 - 9 1304 6200464
U04209.1 - 9 1292

View Full Product Details