Gene Transcripts

Gene Symbol
GPR6
Entrez Gene ID
2830
Gene Name
G protein-coupled receptor 6
Gene Aliases
Chromosome Location
Chr. 6: 109978256 - 109980720 on Build GRCh38
UniGene ID
Hs.46332
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286099.1 NP_001273028.1 3 1757
NM_005284.4 NP_005275.1 2 1648
GenBank mRNA BC043414.2 - 1 1436

View Full Product Details