Gene Transcripts

Gene Symbol
TUSC2
Entrez Gene ID
11334
Gene Name
tumor suppressor candidate 2
Gene Aliases
C3orf11, FUS1, PAP, PDAP2
Chromosome Location
Chr. 3: 50324909 - 50328238 on Build GRCh38
UniGene ID
Hs.517981
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007275.2 NP_009206.1 3 1253
GenBank mRNA AF055479.1 - 3 1261
AK124058.1 - 4 1158
BC023976.2 - 3 1205 3504783

View Full Product Details