Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014470.3 | NP_055285.1 | 5 | 880 | |
GenBank mRNA | AB040147.1 | - | 5 | 858 | |
AK124288.1 | - | 1 | 1368 | ||
AK292762.1 | - | 5 | 880 | ||
AL359573.1 | - | 1 | 113 | ||
BC026356.1 | - | 5 | 880 | 4793442 |