Gene Transcripts
- Gene Symbol
- ZCCHC8
- Entrez Gene ID
- 55596
- Gene Name
- zinc finger CCHC-type containing 8
- Gene Aliases
- Chromosome Location
- Chr. 12: 122471599 - 122501073 on Build GRCh38
- UniGene ID
- Hs.37706
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017612.4 |
NP_060082.2
|
9, 10 |
1102 |
|
XM_006719491.2 |
XP_006719554.1
|
6, 7 |
787 |
|
XM_011538555.2 |
XP_011536857.1
|
8, 9 |
1644 |
|
XM_017019606.1 |
XP_016875095.1
|
9, 10 |
1022 |
|
GenBank mRNA |
AK027702.1 |
- |
9, 10 |
971 |
|
AK074638.1 |
- |
9, 10 |
1000 |
|
AK291174.1 |
- |
5, 6 |
437 |
|
AL157433.1 |
- |
7, 8 |
2511 |
|
BC017704.2 |
- |
2, 3 |
226 |
|
BC065918.1 |
- |
9, 10 |
941 |
5752053 |
View Full Product Details