Gene Transcripts

Gene Symbol
DCHS2
Entrez Gene ID
54798
Gene Name
dachsous cadherin-related 2
Gene Aliases
CDH27, CDHJ, CDHR7, PCDH23, PCDHJ
Chromosome Location
Chr. 4: 154234375 - 154491778 on Build GRCh38
UniGene ID
Hs.655664
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142552.1 NP_001136024.1 4 3011
NM_017639.3 NP_060109.2 8 1144
XM_011532045.1 XP_011530347.1 4 3064
XM_011532046.2 XP_011530348.1 3 720
XM_017008320.1 XP_016863809.1 4 3064
GenBank mRNA AK127704.1 - 4 3003
AY354497.1 - 8 1144
BC140919.1 - 4 2689

View Full Product Details