Gene Transcripts

Gene Symbol
MYO19
Entrez Gene ID
80179
Gene Name
myosin XIX
Gene Aliases
MYOHD1
Chromosome Location
Chr. 17: 36495636 - 36535457 on Build GRCh38
UniGene ID
Hs.302051
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033580.2 NP_001028752.1 6 1356
NM_001163735.1 NP_001157207.1 6 1356
NM_025109.5 NP_079385.2 6 1356
XM_011525286.1 XP_011523588.1 7 905
XM_011525287.1 XP_011523589.1 7 905
XM_011525290.2 XP_011523592.1 7 905
XM_017025157.1 XP_016880646.1 7 879
XM_017025158.1 XP_016880647.1 6 812
XM_017025159.1 XP_016880648.1 7 905
XM_017025160.1 XP_016880649.1 7 905
XM_017025161.1 XP_016880650.1 6 767
GenBank mRNA AB209035.1 - 6 1314
AK026518.1 - 6 767
AK293903.1 - 6 1356
AK304073.1 - 6 707
BC008900.2 - 6 747 3629870

View Full Product Details