Gene Transcripts

Gene Symbol
SYCE1
Entrez Gene ID
93426
Gene Name
synaptonemal complex central element protein 1
Gene Aliases
C10orf94, CT76, POF12, SPGF15
Chromosome Location
Chr. 10: 133553900 - 133568291 on Build GRCh38
UniGene ID
Hs.553795
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143763.1 NP_001137235.1 13 1307
NM_130784.2 NP_570140.1 13 1230
GenBank mRNA AY027807.1 - 13 1230
AY027808.1 - 10 2166

View Full Product Details