Gene Transcripts

Gene Symbol
DPH7
Entrez Gene ID
92715
Gene Name
diphthamide biosynthesis 7
Gene Aliases
C9orf112, RRT2, WDR85
Chromosome Location
Chr. 9: 137554899 - 137578935 on Build GRCh38
UniGene ID
Hs.292570
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138778.3 NP_620133.1 6 843
XM_005266123.2 XP_005266180.1 7 1301
XM_005266126.3 XP_005266183.1 5 1074
XM_005266128.2 XP_005266185.1 7 927
XM_005266129.2 XP_005266186.1 7 930
XM_005266130.2 XP_005266187.1 5 698
XM_011519188.1 XP_011517490.1 6 832
XM_011519189.1 XP_011517491.1 6 944
XM_011519190.2 XP_011517492.1 6 832
XM_011519191.2 XP_011517493.1 6 832
XM_011519192.2 XP_011517494.1 6 832
XM_011519193.2 XP_011517495.1 6 832
XM_011519194.1 XP_011517496.1 6 832
XM_011519199.2 XP_011517501.1 6 789
XM_011519200.1 XP_011517502.1 6 792
XM_017015289.1 XP_016870778.1 6 832
XM_017015290.1 XP_016870779.1 6 832
XM_017015291.1 XP_016870780.1 7 1136
XM_017015292.1 XP_016870781.1 6 1206
XM_017015293.1 XP_016870782.1 6 1163
XM_017015294.1 XP_016870783.1 8 1116
XM_017015295.1 XP_016870784.1 6 1203
XM_017015296.1 XP_016870785.1 6 1042
XM_017015297.1 XP_016870786.1 8 1113
XM_017015298.1 XP_016870787.1 7 1396
XM_017015299.1 XP_016870788.1 7 653
XM_017015300.1 XP_016870789.1 7 1393
XM_017015301.1 XP_016870790.1 5 1066
XM_017015302.1 XP_016870791.1 5 2120
XM_017015303.1 XP_016870792.1 6 1301
XM_017015304.1 XP_016870793.1 5 766
XM_017015305.1 XP_016870794.1 6 2163
XM_017015306.1 XP_016870795.1 7 1301
XM_017015307.1 XP_016870796.1 7 1298
XM_017015308.1 XP_016870797.1 5 2025
XM_017015309.1 XP_016870798.1 7 1025
GenBank mRNA AK075115.1 - 6 844
AK308998.1 - 5 698
AK310226.1 - 2 137
BC003123.1 - 6 782 3502107
BC017335.2 - 6 802 4642600

View Full Product Details