Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_207309.2 | NP_997192.2 | 6 | 1120 | |
XM_006717317.3 | XP_006717380.1 | 6 | 1146 | ||
XM_011519182.2 | XP_011517484.1 | 5 | 714 | ||
GenBank mRNA | AK074143.1 | - | 3 | 1049 | |
AK095393.1 | - | 6 | 861 | ||
AK128529.1 | - | 4 | 955 | ||
AL832421.1 | - | 6 | 889 | ||
BC106035.1 | - | 6 | 832 | ||
BC136391.1 | - | 5 | 687 |