Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001284392.1 | NP_001271321.1 | 4 | 370 | |
NM_001284394.1 | NP_001271323.1 | 5 | 667 | ||
NM_001284398.1 | NP_001271327.1 | 4 | 354 | ||
NM_011129.2 | NP_035259.1 | 5 | 667 | ||
GenBank mRNA | AB078010.1 | - | 4 | 265 | |
AK039859.1 | - | 6 | 677 | ||
AK133933.1 | - | 4 | 322 | ||
AK135143.1 | - | 5 | 668 | ||
AK137299.1 | - | 5 | 586 | ||
BC037673.1 | - | 4 | 372 | ||
BC055101.1 | - | 5 | 667 | 6591016 | |
BU562316.1 | - | 6 | 569 | ||
X61452.1 | - | 5 | 597 |