Gene Transcripts

Gene Symbol
Rab33a
Entrez Gene ID
19337
Gene Name
RAB33A, member RAS oncogene family
Gene Aliases
Rabs10
Chromosome Location
Chr. X: 48519285 - 48530240 on Build GRCm38
UniGene ID
Mm.2015
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011228.2 NP_035358.1 2 1040
GenBank mRNA BC104390.1 - 2 788
BC104391.1 - 2 788 40050296
D83279.1 - 2 855

View Full Product Details