Gene Transcripts

Gene Symbol
Star
Entrez Gene ID
20845
Gene Name
steroidogenic acute regulatory protein
Gene Aliases
AV363654, D8Ertd419e
Chromosome Location
Chr. 8: 25808513 - 25815982 on Build GRCm38
UniGene ID
Mm.293314
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011485.4 NP_035615.2 5 537
GenBank mRNA AK019725.2 - 5 577
AK054470.1 - 5 537
AK136327.1 - 5 547
BC082283.1 - 5 548 30436512
L36062.1 - 5 553

View Full Product Details