Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025283.3 | NP_079559.2 | 8 | 702 | |
GenBank mRNA | AK020446.1 | - | 8 | 658 | |
AK147857.1 | - | 8 | 661 | ||
AK162327.1 | - | 8 | 689 | ||
BC028500.1 | - | 7 | 612 | ||
BC037499.1 | - | 8 | 655 | 1512578 | |
BC052475.1 | - | 8 | 647 | ||
BC058168.1 | - | 8 | 655 | 4980300 | |
U01138.1 | - | 8 | 647 |