Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145584.2 | NP_663559.1 | 16 | 2699 | |
GenBank mRNA | AK146506.1 | - | 16 | 2523 | |
AK173021.1 | - | 13 | 4234 | ||
BC020531.1 | - | 16 | 2569 | 4221758 | |
BC030339.1 | - | 7 | 1153 |