Gene Transcripts
- Gene Symbol
- Ppan
- Entrez Gene ID
- 235036
- Gene Name
- peter pan homolog (Drosophila)
- Gene Aliases
- A230087P06Rik, SSF1
- Chromosome Location
- Chr. 9: 20888175 - 20892179 on Build GRCm38
- UniGene ID
- Mm.2930
- Species
- Mus musculus
- Gene Symbol
- B130055M24Rik
- Entrez Gene ID
- 654803
- Gene Name
- RIKEN cDNA B130055M24 gene
- Gene Aliases
- Chromosome Location
- Chr. 7: 127448654 - 127461238 on Build GRCm38
- UniGene ID
- Mm.423237
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK156871.1 |
- |
|
574 |
|
View Full Product Details