Gene Transcripts

Gene Symbol
Rgs9bp
Entrez Gene ID
243923
Gene Name
regulator of G-protein signalling 9 binding protein
Gene Aliases
R9AP, Rgs9-1bp
Chromosome Location
Chr. 7: 35578993 - 35585582 on Build GRCm38
UniGene ID
Mm.222680
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145840.3 NP_665839.1 1 1308
GenBank mRNA AK044285.1 - 1 1309
AY229888.1 - 1 1270
BC118006.1 - 1 1264 40081388

View Full Product Details