Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001177551.1 | NP_001171022.1 | 2 | 746 | |
NM_153110.4 | NP_694750.3 | 3 | 819 | ||
GenBank mRNA | AK054544.1 | - | 2 | 632 | |
AK136037.1 | - | 2 | 661 | ||
AY063495.1 | - | 2 | 581 | ||
BC064738.1 | - | 3 | 697 | 30049322 |