Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172300.3 | NP_758504.3 | 7 | 1958 | |
GenBank mRNA | AK005524.1 | - | 5 | 1509 | |
AK077793.1 | - | 7 | 1957 | ||
AK149770.1 | - | 7 | 1958 | ||
AK152529.1 | - | 7 | 1672 | ||
AK153328.1 | - | 7 | 1689 | ||
AK168942.1 | - | 7 | 1651 | ||
AK212571.1 | - | 1 | 219 | ||
BC027266.1 | - | 7 | 1583 | ||
BC054412.1 | - | 7 | 1661 | 2646768 |