Gene Transcripts

Gene Symbol
Plxna4
Entrez Gene ID
243743
Gene Name
plexin A4
Gene Aliases
9330117B14, Plxa4, mKIAA1550
Chromosome Location
Chr. 6: 32144543 - 32588192 on Build GRCm38
UniGene ID
Mm.444029
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175750.3 NP_786926.2 2 1844
GenBank mRNA AB073228.1 - 2 1843

View Full Product Details