Gene Transcripts

Gene Symbol
Nlrx1
Entrez Gene ID
270151
Gene Name
NLR family member X1
Gene Aliases
BC034204, NOD9
Chromosome Location
Chr. 9: 44252713 - 44268599 on Build GRCm38
UniGene ID
Mm.3957
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163742.1 NP_001157214.1 6 1206
NM_001163743.1 NP_001157215.1 6 1307
NM_178420.3 NP_848507.2 6 1295
GenBank mRNA AK089260.1 - 6 1299
AK141296.1 - 6 1276
AK145780.1 - 6 1317
AK145988.1 - 6 1296
AK166689.1 - 6 1133
AK170088.1 - 6 1250
BC034204.1 - 5 1089
BC050054.1 - 6 1202 30012053

View Full Product Details