Gene Transcripts
- Gene Symbol
- Notch3
- Entrez Gene ID
- 18131
- Gene Name
- notch 3
- Gene Aliases
- AW229011, N3, hpbk
- Chromosome Location
- Chr. 17: 32120893 - 32166852 on Build GRCm38
- UniGene ID
- Mm.439741
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008716.2 |
NP_032742.1
|
4 |
531 |
|
GenBank mRNA |
X74760.1 |
- |
4 |
531 |
|
View Full Product Details